Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMRT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556106 Supplier Invitrogen™ Supplier No. PA556106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPGPDYNSYK SAYSPSPVEP PSKDFCNFLP TCLDLTMQYS GSGNMELISS NVSVATTYRQ YPLSSRFLVW PKCGPISDTL LYQQCLLNAT TSVQALKPGA SWDLKGARVQ DGLSAEQDMM PSKLEGSLVL PHTPEIQTTR SDLQGHQAVP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.

Gonadal dysgenesis in 9p-deleted individuals might be due to combined hemizygosity of DMRT1 and DMRT2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRT2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DMRT2
Gene Accession No. Q9Y5R5
Gene Alias dmrt2; doublesex and mab-3 related transcription factor 2; doublesex- and mab-3-related transcription factor 2; Doublesex-like 2 protein; doublesex-like 2 protein (DSXL-2); DSXL2; DSXL-2; LOW QUALITY PROTEIN: doublesex- and mab-3-related transcription factor 2; Terra; terra homolog; terra like protein antibody; terra-like protein
Gene Symbols DMRT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DMRT2. Recombinant protein control fragment (Product #RP-100750).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10655
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.