Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMRT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568891
Description
This target displays homology in the following species: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Human: 86%; Mouse: 100%; Rabbit: 86%; Rat: 86%.
Gonadal dysgenesis in 9p-deleted individuals might be due to combined hemizygosity of DMRT1 and DMRT2.
Specifications
DMRT2 | |
Polyclonal | |
Unconjugated | |
DMRT2 | |
dmrt2; doublesex and mab-3 related transcription factor 2; doublesex- and mab-3-related transcription factor 2; Doublesex-like 2 protein; doublesex-like 2 protein (DSXL-2); DSXL2; DSXL-2; LOW QUALITY PROTEIN: doublesex- and mab-3-related transcription factor 2; Terra; terra homolog; terra like protein antibody; terra-like protein | |
Rabbit | |
Protein A | |
RUO | |
226049 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.0 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8BG36 | |
DMRT2 | |
synthetic peptide directed towards the C terminal region of mouse DMRT2. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction