Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMRT3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116427
Description
DMRT3A Polyclonal Antibody for Western Blot
Probable transcription factor that plays a role in configuring the spinal circuits controlling motion in vertebrates. Involved in neuronal specification within specific subdivision of spinal cord neurons and in the development of a coordinated locomotor network controlling movements. May regulate transcription.
Specifications
| DMRT3A | |
| Polyclonal | |
| Unconjugated | |
| dmrt3a | |
| Dmrt3; dmrt3a; dmrt3a1; dmrt7a; doublesex and mab-3 related transcription factor 3; doublesex and mab-3 related transcription factor 3a; doublesex and mab-3 related transcription factor 3a-1; doublesex- and mab-3-related transcription factor 3a; si:dkeyp-31e5.2; zDmart3a; zgc:101766 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 450035 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P83758 | |
| dmrt3a | |
| KLH conjugated synthetic peptide between 341-375 amino acids from the C-terminal region of DANRE dmrt3a. | |
| 200 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction