Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMRTA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566476
Description
Immunogen sequence: VPTLPFRPALD YAFSGMIRDS SYLSSKDSIT CGRLYFRPN Highest antigen sequence identity to the following orthologs - mouse 70%, rat 70%.
DMRTA1 (DMRT Like Family A1) is a Protein Coding gene. Diseases associated with DMRTA1 include Bartholin's Duct Cyst and Cardiomyopathy, Dilated, 1J. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity and sequence-specific DNA binding. An important paralog of this gene is DMRTA2. [GeneCards].
Specifications
DMRTA1 | |
Polyclonal | |
Unconjugated | |
DMRTA1 | |
DMO; DMRT like family A1; DMRT4; Dmrta1; DMRT-like family A1; doublesex and mab-3 related transcription factor 4; doublesex and mab-3 related transcription factor like family A1; doublesex- and mab-3-related transcription factor 4; Doublesex- and mab-3-related transcription factor A1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
63951 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VZB9 | |
DMRTA1 | |
Recombinant Human DMRTA1. Recombinant protein control fragment (Product #RP-107149). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction