Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMRTA1 Polyclonal Antibody

DMRTA1 Polyclonal Antibody
Supplier: Thermo Scientific PA575771
Description
DMRTA1 Polyclonal Antibody for Western Blot
Specifications
| DMRTA1 | |
| Polyclonal | |
| Unconjugated | |
| DMRTA1 | |
| DMO; DMRT like family A1; DMRT4; Dmrta1; DMRT-like family A1; doublesex and mab-3 related transcription factor 4; doublesex and mab-3 related transcription factor like family A1; doublesex- and mab-3-related transcription factor 4; Doublesex- and mab-3-related transcription factor A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 242523, 313352, 63951 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VZB9, Q8CFG4 | |
| DMRTA1 | |
| Synthetic peptide corresponding to residues in Human DMRTA1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction