Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMRTA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535234
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535234 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535234 Supplier Invitrogen™ Supplier No. PA535234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology.

DMRTA2 may be involved in sexual development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRTA2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DMRTA2
Gene Accession No. A2A9A2, Q5UU75, Q96SC8
Gene Alias DMRT like family A2; Dmrt5; Dmrta2; DMRT-like family A2; doublesex and mab-3 related transcription factor like family A2; doublesex- and mab-3-related transcription factor 5; Doublesex- and mab-3-related transcription factor A2
Gene Symbols DMRTA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 487-515 amino acids from the C-terminal region of human DMRTA2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242620, 474350, 63950
Target Species Human, Mouse, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.