Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMRTB1 Monoclonal Antibody (5E7)
GREENER_CHOICE

Catalog No. PIMA524485
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524485 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524485 Supplier Invitrogen™ Supplier No. MA524485
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MPSLAGPPFG AEAAGSGYPG PLDLRRPMRT VPGPLFTDFV RPLNINPDRA LGPEYPGGSS MHPYCPFPLG YLDAPPGVPL QQGFRHVSRS QYQGGGLVSE PGGDFQPSYY LPPPPPPLPP LPPLPPQPQF LPPGYLSALH FLPPPPPPPP PSSFSLTVLF DTDKENTDDQ DAEVLSGEPS QPSSQEQSD.

DMRTB1 gene ontology annotations related to this gene include DNA-binding transcription factor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRTB1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5E7
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene DMRTB1
Gene Accession No. Q96MA1
Gene Alias DMRT like family B with proline rich C-terminal 1; Dmrt6; Dmrtb1; DMRT-like family B with proline-rich C-terminal 1; DMRT-like family B with proline-rich C-terminal, 1; doublesex and mab-3 related transcription factor 6; doublesex- and mab-3-related transcription factor 6; Doublesex- and mab-3-related transcription factor B1; proline-rich protein 13; Prp13; RGD1560921
Gene Symbols DMRTB1
Host Species Mouse
Immunogen DMRTB1 (AAH29566.1, 1 a.a. ~ 189 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63948
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.