Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMRTB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100784
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100784 Supplier Invitrogen™ Supplier No. PA5100784
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DMRTB1.

DMRTB1 gene ontology annotations related to this gene include DNA-binding transcription factor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRTB1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DMRTB1
Gene Accession No. A2A9I7, Q96MA1
Gene Alias DMRT like family B with proline rich C-terminal 1; Dmrt6; Dmrtb1; DMRT-like family B with proline-rich C-terminal 1; DMRT-like family B with proline-rich C-terminal, 1; doublesex and mab-3 related transcription factor 6; doublesex- and mab-3-related transcription factor 6; Doublesex- and mab-3-related transcription factor B1; proline-rich protein 13; Prp13; RGD1560921
Gene Symbols DMRTB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DMRTB1(Accession Q96MA1), corresponding to amino acid residues F294-D342.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56296, 63948
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.