Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMRTB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567812
Description
Predicted to react with Mouse samples.
DMRTB1 gene ontology annotations related to this gene include DNA-binding transcription factor activity.
Specifications
DMRTB1 | |
Polyclonal | |
Unconjugated | |
DMRTB1 | |
DMRT like family B with proline rich C-terminal 1; Dmrt6; Dmrtb1; DMRT-like family B with proline-rich C-terminal 1; DMRT-like family B with proline-rich C-terminal, 1; doublesex and mab-3 related transcription factor 6; doublesex- and mab-3-related transcription factor 6; Doublesex- and mab-3-related transcription factor B1; proline-rich protein 13; Prp13; RGD1560921 | |
Rabbit | |
affinity chromatography | |
RUO | |
56296, 63948 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A2A9I7, Q96MA1 | |
DMRTB1 | |
Synthesized peptide derived from human DMRTB1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction