Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMRTB1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310450100UL
Description
DMRTB1 Polyclonal specifically detects DMRTB1 in Mouse samples. It is validated for Western Blot.Specifications
DMRTB1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
DMRT-like family B with proline-rich C-terminal, 1, doublesex- and mab-3-related transcription factor B1 | |
The immunogen is a synthetic peptide directed towards the c terminal region of mouse DMRTB1 (NP_063925). Peptide sequence PHFLPPGYLSALHFLPPPPPPPSPPSFSLTYDTDKENTNDQDAEAPTEPS | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
63948 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction