Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMRTC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561359 Supplier Invitrogen™ Supplier No. PA561359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLEAAEALLT LRNSAQAPPD SISLHQP Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 56%.

DMRTC1B, also named as Doublesex- and mab-3-related transcription factor C1, is a 192 amino acid protein, which belongs to the DMRT family. DMRTC1B is predominantly expressed in kidney, pancreas, ovary and testis. Detected in brain and in many other tissues. DMRTC1B exists two isoform and molecular weight of isoforms are 20 kDa and 13 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRTC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DMRTC1
Gene Accession No. Q5HYR2
Gene Alias DMRT like family C1; DMRT like family C1B; Dmrt8.2; DMRTC1; DMRTC1B; DMRT-like family C1; DMRT-like family C1B; doublesex and mab-3 related transcription factor 8.2; doublesex- and mab-3-related transcription factor C1; doublesex-mab-3; Gm714
Gene Symbols DMRTC1, DMRTC1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DMRTC1. Recombinant protein control fragment (Product #RP-104411).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63947, 728656
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.