Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMRTC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560896 Supplier Invitrogen™ Supplier No. PA560896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PWVPGHWLPP GFSMPPPVVC RLLYQEPAVS LPPFPGFDPG TSLQLPTHGP FTTCPGSHPV LTAPLSGEPQ GPPSQPRTHS TLILQP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

DMRTC2 belongs to the DMRT family.It may be involved in sexual development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMRTC2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DMRTC2
Gene Accession No. Q8IXT2
Gene Alias 4933432E21Rik; DMRT like family C2; Dmrt7; DMRTC2; DMRT-like family C2; doublesex and mab-3 related transcription factor 7; doublesex and mab-3 related transcription factor like family C2; Doublesex- and mab-3-related transcription factor 7; Doublesex- and mab-3-related transcription factor C2; Unknown (protein for MGC:133756)
Gene Symbols DMRTC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DMRTC2. Recombinant protein control fragment (Product #RP-98459).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63946
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.