Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DMSO, sterile filtered
GSA_VAGREENER_CHOICE

Catalog No. 317625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-762-5ML 25 mL
31-761-00ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 31-762-5ML Supplier Tocris Bioscience™ Supplier No. 3176/25ML
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Solvent for polar and nonpolar compounds

DMSO, sterile filtered is a solvent with wide ranging applications in cell culture, cryopreservation and organic chemistry. DMSO dissolves both polar and nonpolar compounds. Improves responsiveness of human embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) to differentiation signals. DMSO is used in PCR to inhibit secondary structure formation by DNA template or primers.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS

Specifications

Quantity 25 mL
Formula Weight 78.13
Purity >99.9%
Molecular Weight (g/mol) 78.13
Chemical Name or Material DMSO, sterile filtered
Recommended Storage Store at RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.