Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMTF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540935
Description
Peptide sequence: SFNDAHVSKF SDQNSTELMN SVMVRTEEEI SDTDLKQEES PSDLASAYVT Sequence homology: Dog: 92%; Guinea Pig: 91%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 92%.
This gene encodes a transcription factor that contains a cyclin D-binding domain, three central Myb-like repeats, and two flanking acidic transactivation domains at the N- and C-termini. The encoded protein is induced by the oncogenic Ras signaling pathway and functions as a tumor suppressor by activating the transcription of ARF and thus the ARF-p53 pathway to arrest cell growth or induce apoptosis. It also activates the transcription of aminopeptidase N and may play a role in hematopoietic cell differentiation. The transcriptional activity of this protein is regulated by binding of D-cyclins. This gene is hemizygously deleted in approximately 40% of human non-small-cell lung cancer and is a potential prognostic and gene-therapy target for non-small-cell lung cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
DMTF1 | |
Polyclonal | |
Unconjugated | |
DMTF1 | |
cyclin D binding myb like transcription factor 1; cyclin D binding myb-like transcription factor 1; cyclin D binding myb-like transcription factor 1D-interacting myb-like protein; cyclin D-binding Myb-like protein; cyclin-D-binding Myb-like transcription factor 1; Cyclin-D-interacting Myb-like protein 1; Dimp; D-interacting myb-like protein; Dmp1; DMTF; Dmtf1; FLJ41265; hDMP1; hDMTF1; mDmp1; MRUL | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9988 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y222 | |
DMTF1 | |
Synthetic peptide directed towards the C-terminal of human DMTF1 (aa 626-675). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction