Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMTF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582848 Supplier Invitrogen™ Supplier No. PA582848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASPTVTLTAA APASPEQIIV HALSPEHLLN TSDNVTVQCH TPRVIIQTVA TEDITSSISQ AELTVDSDIQ SSDFPEPPDA LEADTFPDEI HHPKMTVEPS FNDAHVSKFS DQNSTELMNS VMVRTEEEI Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 91%.

This gene encodes a transcription factor that contains a cyclin D-binding domain, three central Myb-like repeats, and two flanking acidic transactivation domains at the N- and C-termini. The encoded protein is induced by the oncogenic Ras signaling pathway and functions as a tumor suppressor by activating the transcription of ARF and thus the ARF-p53 pathway to arrest cell growth or induce apoptosis. It also activates the transcription of aminopeptidase N and may play a role in hematopoietic cell differentiation. The transcriptional activity of this protein is regulated by binding of D-cyclins. This gene is hemizygously deleted in approximately 40% of human non-small-cell lung cancer and is a potential prognostic and gene-therapy target for non-small-cell lung cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMTF1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DMTF1
Gene Accession No. Q9Y222
Gene Alias cyclin D binding myb like transcription factor 1; cyclin D binding myb-like transcription factor 1; cyclin D binding myb-like transcription factor 1D-interacting myb-like protein; cyclin D-binding Myb-like protein; cyclin-D-binding Myb-like transcription factor 1; Cyclin-D-interacting Myb-like protein 1; Dimp; D-interacting myb-like protein; Dmp1; DMTF; Dmtf1; FLJ41265; hDMP1; hDMTF1; mDmp1; MRUL
Gene Symbols DMTF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DMTF1. Recombinant protein control fragment (Product #RP-89350).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9988
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.