Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMWD Monoclonal Antibody (3F5)
GREENER_CHOICE

Catalog No. PIMA518749
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518749 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518749 Supplier Invitrogen™ Supplier No. MA518749
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: GGEKPSGPVP RSRLDPAKVL GTALCPRIHE VPLLEPLVCK KIAQERLTVL LFLEDCIITA CQEGLICTWA RPGKAGISSQ PGNSPSGTVV.

DMWD (DM1 Locus, WD Repeat Containing) is a Protein Coding gene. Diseases associated with DMWD include Myotonic Dystrophy 1 and Corticosteroid Allergy. An important paralog of this gene is WDR20. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMWD
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F5
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Dmwd
Gene Accession No. Q09019
Gene Alias D19S593E; Dm9; DMRN9; DMR-N9; DMWD; dystrophia myotonica linked gene, N9; dystrophia myotonica WD repeat-containing protein; dystrophia myotonica, WD repeat containing; dystrophia myotonica-containing WD repeat motif; dystrophia myotonica-containing WD repeat motif protein; gene59; Protein 59; Protein DMR-N9
Gene Symbols Dmwd
Host Species Mouse
Immunogen DMWD (AAH19266, 245 a.a. ~ 334 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1762
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.