Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMWD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564432 Supplier Invitrogen™ Supplier No. PA564432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TYLKWLPESE SLFLASHASG HLYLYNVSHP CASAPPQYSL LKQGEGFSVY A Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.

DMWD (DM1 Locus, WD Repeat Containing) is a Protein Coding gene. Diseases associated with DMWD include Myotonic Dystrophy 1 and Corticosteroid Allergy. An important paralog of this gene is WDR20. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMWD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dmwd
Gene Accession No. Q09019
Gene Alias D19S593E; Dm9; DMRN9; DMR-N9; DMWD; dystrophia myotonica linked gene, N9; dystrophia myotonica WD repeat-containing protein; dystrophia myotonica, WD repeat containing; dystrophia myotonica-containing WD repeat motif; dystrophia myotonica-containing WD repeat motif protein; gene59; Protein 59; Protein DMR-N9
Gene Symbols Dmwd
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DMWD. Recombinant protein control fragment (Product #RP-103941).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1762
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.