Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMWD Rabbit anti-Human, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 160701AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-0690


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Tight junctions form an important barrier of paracellular transport in epithelial cells. Sealing of two adjacent cells at bicellular tight junctions, a point where three adjacent cells are in contact with each other. Tricellulin is the first protein identified that specifically concentrates in tricellular tight junctions. This protein has four membrane spanning domains, similarly to claudins. Tricellulin expression is high in epithelium-derived tissues, such as small intestine, kidney and lung. Functional evidence for the role of tricellulin in tight junction formation comes from siRNA studies, where suppression of its expression leads to compromised epithelial barrier and tight junction formation. Loss of function in tricellulin mutants missing all or most of a conserved region in the cytosolic domain which binds to the cytosolic scaffolding protein ZO-1, indicate that interaction with other known tight junction proteins plays an important role for the function of tricellulin.
Specifications

Specifications

DMWD
Polyclonal
Unconjugated
DMWD
D19S593E, DM9, DMR N9, DMRN9, DMWD, gene59, Protein 59, Protein DMR N9
Rabbit
Antigen Affinity Chromatography
RUO
1762, 680252
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q09019
DMWD
DMWD Fusion Protein Ag9007
150 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.