Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DMXAA
GREENER_CHOICE

Catalog No. 560110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
560110 10 mg
560150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 560110 Supplier Tocris Bioscience™ Supplier No. 5601/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

mSTING agonist; induces antitumor immunological responses

DMXAA is a mSTING agonist; selective for mouse STING over human STING. Induces IFN-β and cytokine production from bone marrow-derived dendritic cells. Causes tumor regression following intratumoral administration in mouse tumor models. Induces an adaptive immune response to reduce systemic tumor growth and provides immunological memory against autologous tumor re-challenge. Also antiviral.

Specifications

Chemical Name or Material 5,6-Dimethyl-9-oxo-9 H-xanthene-4-acetic acid
CAS 117570-53-3
Quantity 10 mg
Target STING Agonists
Molecular Formula C17H14O4
Formula Weight 282.29
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.