Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DMXL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557655 Supplier Invitrogen™ Supplier No. PA557655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDVSGILATQ VYTWVDDDIE VETKGSEDFL VIHARDDLTA VQGTTPYTHS NPGTPINMPW LGSTQTGRGA SVMIKKAINN VRRMTSHPTL P Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.

The protein encoded by this gene is a member of the WD repeat superfamily of proteins, which have regulatory functions. This gene is expressed in many tissue types including several types of eye tissue, and it has been associated with ocular phenotypes. In addition, it is upregulated in cultured cells that overexpress growth factor independence 1B, a transcription factor that is essential for hematopoietic cell development. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DMXL1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DMXL1
Gene Accession No. Q9Y485
Gene Alias Dmx like 1; DMXL1; Dmx-like 1; dmX-like protein 1; XL1; X-like 1 protein
Gene Symbols DMXL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DMXL1. Recombinant protein control fragment (Product #RP-96940).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1657
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.