Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DMXL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558762
Description
Immunogen sequence: TKTSALSAKK DQPDFISHRM DDVPSHSKAL SDGNGSSGIE WSNVTSSQYD WSQPIVKVDE EPLNLDWGED HDSALDEEED DAVGLVMKST DA Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 67%.
This gene encodes a protein with 12 WD domains. Proteins with WD domains are involved in many functions including participation in signal transduction pathways. Participation of the encoded protein in regulation of the Notch signaling pathway has been demonstrated in vitro using several human cell lines. A gene encoding a similar protein is located on chromosome 5. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
DMXL2 | |
Polyclonal | |
Unconjugated | |
DMXL2 | |
6330586A12; 6430411K14Rik; Dmx like 2; DMXL2; Dmx-like 2; dmX-like protein 2; E130119P06Rik; KIAA0856; mKIAA0856; PEPNS; rabconnectin 3; rabconnectin-3; RC3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23312 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TDJ6 | |
DMXL2 | |
Recombinant protein corresponding to Human DMXL2. Recombinant protein control fragment (Product #RP-98044). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction