Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Antibody (SPM603), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311432AF488
Description
DNA Monoclonal antibody specifically detects DNA in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DNA | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SPM603 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Double stranded deoxyribonucleic acid, Double stranded DNA, dsDNA | |
Nuclei of Burkitt's cells | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction