Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Bulletin Board Set
SDPSCI_ED

Catalog No. S02419
Click to view available options
Product Type:
Habitat Bulletin Board Set
Plants Bulletin Board Set
Grade:
4 to 8

An informative and anatomy-themed bulletin board set.

Specifications

Grade 4 to 8
Format Set
Product Type Habitat Bulletin Board Set
Includes 5 Charts that feature detailed artwork, resource guide
For Use With (Application) An informative nature themed bulletin board
Subject Biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.