Description
- DNA DuraGel™ gels are permanent polymer gels that allow students to practice the critically important skill of pipetting/gel loading.
- The clear, reusable gels are designed for the practice of loading 5 - 35 μl of samples.
- The gel grids are imprinted with a ruler for sizing DNA fragments.Also included are simulated FlashBlue™ and Ethidium Bromide gel images, ideal for representing how actual gels are stained with Methylene Blue and Ethidium Bromide.
Specifications
Specifications
| Device Holds | 6 Gels (4 to 8 Students) |
| Type | DNA DuraGel™ |
| Includes | 2 Reusable DNA DuraGel™, 2 FlashBlue™ and 2 Ethidium Bromide gel images, practice gel loading solution and mini-transfer pipets |