Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA G-quadruplex (G4) Mouse anti-All, Clone: 1H6, MilliporeSigma™

Catalog No. MABE1126MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1126MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1126MI Supplier MilliporeSigma Supplier No. MABE1126
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-DNA G-quadruplex (G4) Antibody, clone 1H6 is validated for use in Immunocytochemistry, Immunohistochemistry (Paraffin), ELISA and Flow Cytometry for the detection of DNA G-quadruplex .
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA G-quadruplex (G4)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), ELISA, Flow Cytometry
Classification Monoclonal
Clone 1H6
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Mouse
Immunogen KLH-conjugated G-quadruplex DNA derived from oligonucleotides with telomeric repeats.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 1H6 binds both tetramolecular and unimolecular (intramolecular) DNA G-quadruplex (G4) structures without sequence specificity, while exhibiting significantly lower affinity toward tetramolecular RNA structure or a triplex DNA structure, and little to no affinity toward either non-G4 ssDNA or ddDNA. However, clone 1H6 does not seem to bind the [AGGG(TTAGGG)3] unimolecular G4 structure, indicating a broad selectivity toward many, but not all, DNA G4 structures (Henderson, A., et al. (2014). Nucleic Acids Res. 42(2):860-869).
Target Species All species
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.