Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Helicase B (HELB) Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE2600100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2600-100 100 μL
ABE2600-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE2600-100 Supplier MilliporeSigma Supplier No. ABE2600100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-DNA Helicase B (HELB) Antibody is a highly specific rabbit polyclonal antibody that targets DNA helicase B (HELB) and has been tested for use in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Helicase B (HELB)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Helb
Host Species Rabbit
Immunogen GST-tagged recombinant fragment corresponding to 227 amino acids from the C-terminal half of murine DNA helicase B.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_536694
Test Specificity This rabbit polyclonal antibody detects DNA helicase B in murine species. It targets an epitope within 227 amino acids from the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.