Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Ligase I Antibody (1A9), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308267AF532
Description
DNA Ligase I Monoclonal antibody specifically detects DNA Ligase I in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
DNA Ligase I | |
Monoclonal | |
Alexa Fluor 532 | |
DNA ligase 1, DNA ligase I, EC 6.5.1.1, ligase I, DNA, ATP-dependent, MGC117397, MGC130025, Polydeoxyribonucleotide synthase [ATP] 1, polydeoxyribonucleotide synthase ATP 1 | |
Full-length native bovine DNA Ligase I protein (Uniprot: P18858) | |
100 μg | |
Apoptosis, Base Excision Repair, Cancer, DNA Repair, Nucleotide Excision Repair, Tumor Suppressors | |
3978 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunoprecipitation | |
1A9 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction