Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA Ligase I Monoclonal Antibody (10H5)
GREENER_CHOICE

Catalog No. PIMA123189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA123189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA123189 Supplier Invitrogen™ Supplier No. MA123189
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: MCF7.

The protein encoded by this gene is involved in the synthesis of asparagine. This gene complements a mutation in the temperature-sensitive hamster mutant ts11, which blocks progression through the G1 phase of the cell cycle at nonpermissive temperature. There are three alternatively spliced transcript variants encoding the same protein described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Ligase I
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10H5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LIG1
Gene Accession No. P18858, P37913
Gene Alias AL033288; DNA ligase (ATP) 1; DNA ligase 1; DNA ligase I; LIG1; Lig-1; ligase 1; ligase I, DNA, ATP-dependent; LigI; MGC117397; MGC130025; polydeoxyribonucleotide synthase [ATP] 1
Gene Symbols LIG1
Host Species Mouse
Immunogen Full-length recombinant human DNA Ligase I protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16881, 3978
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.