Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNA Ligase I Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA129815
Description
PA1-29815 detects Ligase 1 from human samples.
The protein encoded by this gene is involved in the synthesis of asparagine. This gene complements a mutation in the temperature-sensitive hamster mutant ts11, which blocks progression through the G1 phase of the cell cycle at nonpermissive temperature. There are three alternatively spliced transcript variants encoding the same protein described for this gene.
Specifications
DNA Ligase I | |
Polyclonal | |
Unconjugated | |
LIG1 | |
AL033288; DNA ligase (ATP) 1; DNA ligase 1; DNA ligase I; LIG1; Lig-1; ligase 1; ligase I, DNA, ATP-dependent; LigI; MGC117397; MGC130025; polydeoxyribonucleotide synthase [ATP] 1 | |
Rabbit | |
Protein G | |
RUO | |
3978 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.1mg/mL BSA and 0.02% sodium azide | |
P18858 | |
LIG1 | |
Synthetic peptide, corresponding to amino acids 51-68 (SPVKRPGRKAARVLGSEG) of Human Lig1. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction