Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA Ligase I Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595963 Supplier Invitrogen™ Supplier No. PA595963
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CEYKYDGQRA QIHALEGGEV KIFSRNQEDN TGKYPDIISR IPKIKLPSVT SFILDTEAVA WDREKKQIQP FQVLTTRKRK EVDASEIQVQ VCLYAFDLIY LNGESLVREP LSRRRQLLRE NFVETEGEFV FATSLDTKDI EQIAEFLEQS VKDSCEGLMV KTLDVDATYE IAKRSHNWLK LKKDYLDGVG DTLDLVVIGA YLGRGKRAGR YGGFLLASYD EDSEEL; Positive Samples: MCF7, A-549, K-562, Jurkat, 22Rv1, Mouse testis; Cellular Location: Nucleus.

The protein encoded by this gene is involved in the synthesis of asparagine. This gene complements a mutation in the temperature-sensitive hamster mutant ts11, which blocks progression through the G1 phase of the cell cycle at nonpermissive temperature. There are three alternatively spliced transcript variants encoding the same protein described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Ligase I
Applications ELISA, Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LIG1
Gene Accession No. P18858, P37913
Gene Alias AL033288; DNA ligase (ATP) 1; DNA ligase 1; DNA ligase I; LIG1; Lig-1; ligase 1; ligase I, DNA, ATP-dependent; LigI; MGC117397; MGC130025; polydeoxyribonucleotide synthase [ATP] 1
Gene Symbols LIG1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 565-790 of human LIG1 (NP_0002251).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16881, 3978
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.