Learn More
Invitrogen™ DNA Ligase I Recombinant Rabbit Monoclonal Antibody (JB43-39)

Description
Positive Control: Daudi, A431, MCF-7, Rat testis tissue, human tonsil tissue, human spleen tissue.
Specifications
Specifications
| Antigen | DNA Ligase I |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | JB43-39 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | LIG1 |
| Gene Accession No. | P18858 |
| Gene Alias | AL033288; DNA ligase (ATP) 1; DNA ligase 1; DNA ligase I; LIG1; Lig-1; ligase 1; ligase I, DNA, ATP-dependent; LigI; MGC117397; MGC130025; polydeoxyribonucleotide synthase [ATP] 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.