Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA Ligase III Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570056 Supplier Invitrogen™ Supplier No. PA570056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA Ligase III gene product migrates as a 100 kDa (a-form) protein in somatic tissues, is ubiquitously expressed and complexes with XRCC-1 and a 97 kDa (beta form) protein that is found in testis. DNA Ligase III is involved in rejoining DNA single-strand breaks and in the base excision repair pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Ligase III
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LIG3
Gene Accession No. P97386
Gene Alias D11Wsu78e; DNA ligase (ATP) 3; DNA ligase 3; DNA ligase III; hypothetical protein; LIG2; LIG3; ligase II, DNA, ATP-dependent; ligase III, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 3; Unknown (protein for MGC:134139)
Gene Symbols LIG3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human DNA Ligase III (aa 933-982).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16882
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.