Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNA Ligase III Recombinant Rabbit Monoclonal Antibody (5I7T5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551407
Description
Immunogen sequence: IASRCTANDL KCIIRLIKHD LKMNSGAKHV LDALDPNAYE AFKASRNLQD VVERVLHNAQ EVEKEPGQRR ALSVQASLMT PVQPMLAEAC KSVEYAMKKC P.
There are four distinct ligase proteins found in mammalian cells. The activity of these DNA ligases is required for the joining of DNA strand breaks that occur during DNA replication, repair and recombination. DNA Ligase III gene product migrates as a 100 kDa (a-form) protein in somatic tissues, is ubiquitously expressed and complexes with XRCC-1 and a 97 kDa (beta form) protein that is found in testis. DNA Ligase III is involved in rejoining DNA single-strand breaks and in the base excision repair pathway.
Specifications
DNA Ligase III | |
Recombinant Monoclonal | |
0.09 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P49916, P97386 | |
LIG3 | |
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human DNA Ligase III/LIG3 (NP_039269.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5I7T5 | |
Unconjugated | |
LIG3 | |
D11Wsu78e; DNA ligase (ATP) 3; DNA ligase 3; DNA ligase III; hypothetical protein; LIG2; LIG3; ligase II, DNA, ATP-dependent; ligase III, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 3; Unknown (protein for MGC:134139) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16882, 303369, 3980 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction