Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA Ligase IV Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540826
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540826 Supplier Invitrogen™ Supplier No. PA540826
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DGERMQMHKD GDVYKYFSRN GYNYTDQFGA SPTEGSLTPF IHNAFKADIQ Sequence homology: Cow: 92%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 93%.

The protein encoded by this gene is a DNA ligase that joins single-strand breaks in a double-stranded polydeoxynucleotide in an ATP-dependent reaction. This protein is essential for V(D)J recombination and DNA double-strand break (DSB) repair through nonhomologous end joining (NHEJ). This protein forms a complex with the X-ray repair cross complementing protein 4 (XRCC4), and further interacts with the DNA-dependent protein kinase (DNA-PK). Both XRCC4 and DNA-PK are known to be required for NHEJ. The crystal structure of the complex formed by this protein and XRCC4 has been resolved. Defects in this gene are the cause of LIG4 syndrome. Alternatively spliced transcript variants encoding the same protein have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA Ligase IV
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LIG4
Gene Accession No. P49917
Gene Alias 5830471N16Rik; DNA joinase; DNA ligase (ATP) 4; DNA ligase 4; DNA ligase IV; DNA repair enzyme; Lig4; LIG4S; ligase IV, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 4; polynucleotide ligase; sealase; tiny
Gene Symbols LIG4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LIG4 (aa 275-324).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3981
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.