Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Ligase IV Rabbit anti-Human, Clone: S08-3B4, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314926100UL
This item is not returnable.
View return policy
Description
DNA Ligase IV Monoclonal antibody specifically detects DNA Ligase IV in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
DNA Ligase IV | |
Monoclonal | |
Unconjugated | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
S08-3B4 | |
Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500 | |
DNA joinase, DNA ligase 4, DNA ligase IV, DNA repair enzyme, EC 6.5.1.1, ligase IV, DNA, ATP-dependent, Polydeoxyribonucleotide synthase [ATP] 4, polynucleotide ligase, sealase | |
Recombinant protein of human DNA Ligase IV (Uniprot#: P49917) | |
100 μg | |
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining | |
3981 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction