Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ DNA LoBind™ Tubes
GSA_VA

Catalog No. 13698792 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Max. RCF:
25,000 x g
30,000 x g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Max. RCF
13-698-792 2 mL 25,000 x g
13-698-791 1.5 mL 30,000 x g
13-698-790 0.5 mL 30,000 x g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-698-792 Supplier Eppendorf™ Supplier No. 022431048
Get More Eppendorf Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unleash the power of precision in your DNA research with Eppendorf DNA LoBind Tubes. Engineered with proprietary LoBind technology, these microcentrifuge tubes redefine sample handling, minimizing the risk of loss and maximizing DNA recovery. Ideal for low-concentration applications, including PCR, qPCR, and DNA sequencing, these tubes ensure accuracy, purity, and optimal yields.

  • Surface-coating-free, including substances like silicone, plasticizers, biocides or mold release agents to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Precision lid sealing minimizes evaporation rates in the tube format.
  • Suitable for molecular biology applications with a centrifugation speed rating of up to 30,000 x g (25,000 x g for 2.0 mL tube).
  • Snap, screw, and SnapTec™ cap options available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DNA, DNase, RNase, PCR Inhibitors free
Closure Material HDPE
Color Clear
Capacity (Metric) 2 mL
Material Polypropylene
For Use With DNA or RNA applications in forensic analysis, microarrays, next generation sequencing, gene cloning and general molecular biology
Includes Safe-lock lid
No. per Pack 250 Pk.
Packaging 250 Tubes (5 Bags x 50 Tubes)
Sterility Non-sterile
RNase Free Yes
Dnase Free Yes
Autoclavable Autoclavable
Shape Cylindrical
Capacity (English) 0.07 oz.
Clean Claims PCR Inhibitor Free
Bottom Shape Conical
Closure Type Snap Cap
Graduated Yes
Product Type Microcentrifuge Tube
Max. RCF 25,000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.