Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA Monoclonal Antibody (49/4A1)
GREENER_CHOICE

Catalog No. MA183116
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA183116 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA183116 Supplier Invitrogen™ Supplier No. MA183116
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Bovine DNA antibody, clone ET844.2 recognizes both double and single stranded eukaryotic DNA.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA
Applications ELISA, Immunodiffusion
Classification Monoclonal
Clone 49/4A1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide
Gene Alias dsdna; e. coli dna; single-stranded DNA
Host Species Mouse
Immunogen DNA from calf thymus.
Purification Method PEG precipitation
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.