Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNA Monoclonal Antibody (ET844.2), Invitrogen™

Catalog No. PIMA545997
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545997 1 mg
PIMA546033 100 μg
01-671-553 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIMA545997 Supplier Invitrogen™ Supplier No. MA545997
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes single- or double-stranded eucaryotic DNA as well as E. coli DNA. Reactivity with other prokaryotic DNA has not been investigated.

DNA Monoclonal antibody specifically detects DNA in Many samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA
Applications ELISA
Classification Monoclonal
Clone ET844.2
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias dsdna; e. coli dna; single-stranded DNA
Host Species Mouse
Immunogen Calf thymus DNA.
Purification Method PEG precipitation
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.