Learn More
Promega DNA-PK Kinase Enzyme System, 2,500 units
Catalog No. PRV4106 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
2500 Units
Catalog No. Quantity
PRV4106 2500 Units
1 options
Catalog No. PRV4106 Supplier Promega Supplier No. V4106
Add to Cart
Add to Cart
Native human DNA-Dependent Protein Kinase purified from HeLa cells. Consists of a ∼460kDa catalytic subunit and a heterodimeric DNA-binding subunit (Ku) containing an 85kDa and a 70kDa peptide (1). Gene sequence can be found at accession number NM_006904.
  • Easily Screen and Profile DNA−PK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 2500 Units
Content And Storage <-65°C