Learn More
Invitrogen™ DNA-PK Monoclonal Antibody (3H6)

Description
MA5-15813 targets PRKDC in indirect ELISA, ICC, IHC, IF and WB applications and shows reactivity with Human samples. The MA5-15813 immunogen is purified recombinant fragment of human PRKDC expressed in E. Coli. MA5-15813 detects PRKDC which has a predicted molecular weight of approximately 46.9kDa.
Specifications
Specifications
| Antigen | DNA-PK |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3H6 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | PRKDC |
| Gene Accession No. | P78527 |
| Gene Alias | AI326420; AU019811; DNA-dependent protein kinase catalytic subunit; DNAPDcs; DNAPK; DNA-PK catalytic subunit; DNA-PKcs; DNPK1; DOXNPH; doxorubicin nephropathy; dxnph; hyper-radiosensitivity of murine scid mutation, complementing 1; HYRC; HYRC1; I79_017613; IMD26; p350; p460; Prkdc; protein kinase, DNA activated, catalytic polypeptide; protein kinase, DNA-activated, catalytic polypeptide; scid; severe combined immunodeficiency; slip; Xrcc7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.