Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA-PKcs Ab-1 Monoclonal Antibody, Epredia™

Catalog No. MS369P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS369P 1 mL Unconjugated Concentrated 200μg/mL
MS369B0 100 μL Biotin Concentrated 200μg/mL
MS369B1 500 μL Biotin Concentrated 200μg/mL
MS369B 1 mL Biotin Concentrated 200μg/mL
MS369P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS369PABX 200 μL Unconjugated Concentrated 1mg/mL
MS369R7 7 mL Unconjugated Prediluted Prediluted
MS369P0 100 μL Unconjugated Concentrated 200μg/mL
MS369P1 500 μL Unconjugated Concentrated 200μg/mL
MS369R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS369P Supplier Epredia Supplier No. MS369P
Add to Cart
Add to Cart

Inhibit DNA-dependent phosphorylation and perform other studies with Epredia™ DNA-PKcs Ab-1, Mouse Monoclonal Antibody.

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 18-2

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 1-2713

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: LS174T cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Inhibits (50%) DNA-dependent Phosphorylation

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen DNA-PKcs Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 18-2
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human DNA-dependent Protein Kinase purified from HeLa cells
Quantity 1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.