Learn More
Description
Host Species: Mouse
Clone: 18-2
Isotype: IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: aa 1-2713
Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells
Molecular Weight: 460 kDa
Positive Control: LS174T cells, Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Inhibits (50%) DNA-dependent Phosphorylation
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | DNA-PKcs Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 18-2 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human DNA-dependent Protein Kinase purified from HeLa cells |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.