Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA-PKcs Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100658
Description
DNA-PKcs Polyclonal specifically detects DNA-PKcs in Human samples. It is validated for Western Blot, Immunoprecipitation, Knockdown Validated.Specifications
DNA-PKcs | |
Polyclonal | |
Unconjugated | |
TBS, 0.1% BSA with 0.09% Sodium Azide | |
DNA-dependent protein kinase, DNA-dependent protein kinase catalytic subunit-like, DNA-PK catalytic subunit, DNA-PKcs, DNPK1, EC 2.7.11.1, HYRC, HYRC1, p460 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunoprecipitation, KnockDown | |
0.2 mg/mL | |
Western Blot 1:1000 - 1:10000, Immunoprecipitation 2 - 5 ug/mg lysate, KnockDown Validated | |
P78527 | |
PRKDC | |
The immunogen recognized by this antibody maps to a region between residues 2050 and 2100 of human DNA-Dependent Protein Kinase, catalytic subunit using the numbering given in entry NP_008835.5 (GeneID 5591). | |
0.1 mL | |
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining, Protein Kinase | |
5591 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction