Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ DNA-PKcs, Mouse, Unlabeled, Clone: 3A9.1,

Catalog No. MABC1236MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC1236MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1236MI Supplier MilliporeSigma™ Supplier No. MABC1236
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA-dependent protein kinase catalytic subunit (EC 2.7.11.1; UniProt P78527; also known as DNA-PK catalytic subunit, DNA-PKcs, DNPK1, p460) is encoded by the PRKDC (also known as HYRC, HYRC1, IMD26, XRCC7) gene (Gene ID 5591) in human. DNA-PK is a heterotrimer of PRKDC and the Ku p70/YRCC6-p86/XRCC5 dimer. It acts as a molecular sensor for DNA damage and is involved in DNA non-homologous end joining required for double-strand break (DSB) repair and V(D)J recombination. Upon binding to DNA, it expresses its catalytic properties. DNA-PKcs falls into the phosphatidylinositol (PI) 3-kinase family, however, biochemical assays indicate that it phosphorylates proteins and has no detectable activity toward lipids. DNA-PKcs is displays high similarity to PI kinase family members involved in cell cycle control, DNA repair, and DNA damage responses. Deficiency of DNA-PKcs is linked to severe combined immunodeficiency characterized by reduced or absent T and B cells, recurrent candidiasis, and lower respiratory tract infections. Ref.: Hartley, KO., et al. (1995). 82, 849-856.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNA-PKcs
Applications Western Blot
Classification Monoclonal
Clone 3A9.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene PRKDC, HYRC, HYRC1, IMD26, XRCC7
Gene Accession No. P78527
Host Species Mouse
Immunogen GST-tagged recombinant fragment from the region between FAT and KIP-binding domain of human DNA-PKcs
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_008835
Test Specificity Clone 3A9.1 detects DNA-PK catalytic subunit (DNA-PKcs), but not the regulatory subunit (Ku). Target epitope is present in both spliced isoforms of human DNA-PKcs reported by UniProt (P78527).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.