Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Polymerase beta Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305939JF646
Description
DNA Polymerase beta Polyclonal specifically detects DNA Polymerase beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DNA Polymerase beta | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DNA pol beta, DNA polymerase beta, DNA polymerase beta subunit, EC 2.7.7.7, EC 4.2.99.-, MGC125976, polymerase (DNA directed), beta | |
Produced in rabbits immunized with E. coli-derived Human DNA Polymerase beta fragment. | |
0.1 mL | |
DNA Polymerases, DNA Repair, DNA replication Transcription Translation and Splicing | |
5423 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction