Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase beta Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988620
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8620 Supplier Bioss Supplier No. BS8525R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA polymerase beta comprises an amino-terminal 8- kDa domain and a carboxy-terminal 31- kDa domain. The N-terminal ssDNA binding domain has a deoxyribose phosphodiesterase activity while the C-terminal domain has a nucleotidyltransferase activity. Mammalian DNA polymerase beta, a DNA repair polymerase, is constitutively expressed in cultured cells, but treatment of cells with the DNA-alkylating agents such as N-methyl-N′-nitro-N-nitrosoguanidine (MNNG) or methyl methanesulfonate up-regulates beta-pol level. DNA polymerase beta fills single nucleotide gaps in DNA produced by the base excision repair pathway of mammalian cells.

Specifications

Antigen DNA Polymerase beta
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene POLB
Gene Accession No. P06746
Gene Alias A430088C08Rik; BOS_23903; DNA pol beta; DNA polymerase beta; DNA polymerase beta subunit; DNA-directed DNA polymerase beta; LOW QUALITY PROTEIN: DNA polymerase beta; MGC125976; POL B; Pol beta; POLB; polb.S; polymerase (DNA directed), beta; polymerase (DNA directed), beta S homeolog; polymerase (DNA) beta; polymerase B; polymerase-beta; wu:fa97a05; XELAEV_18020165mg; zgc:109924
Gene Symbols POLB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DNA Polymerase beta.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5423
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.