Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Polymerase delta, catalytic subunit Rabbit anti-Human, Mouse, Rat, Clone: 4B6I3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | DNA Polymerase delta, catalytic subunit |
---|---|
Clone | 4B6I3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
DNA Polymerase delta, catalytic subunit Monoclonal antibody specifically detects DNA Polymerase delta, catalytic subunit in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
DNA Polymerase delta, catalytic subunit | |
Western Blot | |
Unconjugated | |
Rabbit | |
Breast Cancer, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5424 | |
IgG | |
Affinity purified |
4B6I3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CDC2, CDC2 homolog, DNA polymerase delta catalytic subunit, DNA polymerase subunit delta p125, DNA-directed polymerase delta 1, EC 2.7.7.7, POLD, polymerase (DNA directed), delta 1, catalytic subunit (125kD), polymerase (DNA directed), delta 1, catalytic subunit 125kDa | |
A synthetic peptide corresponding to a sequence within amino acids 1000-1107 of human DNA Polymerase delta, catalytic subunit (P28340). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title