Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA polymerase delta p50 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232093IR
Description
DNA polymerase delta p50 Polyclonal antibody specifically detects DNA polymerase delta p50 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
DNA polymerase delta p50 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
delta 2, regulatory subunit (50kD), polymerase (DNA directed), delta 2, regulatory subunit 50kDa | |
The immunogen this antibody was made to, maps to a region between residue 419 to 469 of human Polymerase (DNA directed), Delta 2, Accessory Subunit using the numbering given in entry NP_001120690.1 (GeneID 5425). | |
0.1 mL | |
Cell Cycle and Replication | |
5425 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction