Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Polymerase epsilon catalytic subunit A Antibody (3C5.1) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-4976870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100115SS 0.025 mg
NB100115 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100115SS Supplier Novus Biologicals Supplier No. NB100115SS

Mouse Monoclonal Antibody has been used in 9 publications

DNA Polymerase epsilon catalytic subunit A Monoclonal specifically detects DNA Polymerase epsilon catalytic subunit A in Human, Mouse, Mustelid, Primate, Drosophila (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen DNA Polymerase epsilon catalytic subunit A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 3C5.1
Concentration 16 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 29935943), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200
Gene Accession No. Q9NRF9
Gene Alias DKFZp434F222, DNA polymerase epsilon catalytic subunit A, DNA polymerase II subunit A, EC 2.7.7, EC 2.7.7.7, POLE1FLJ21434, polymerase (DNA directed), epsilon
Gene Symbols POLE
Host Species Mouse
Immunogen Human DNA Polymerase epsilon purified from HeLa cells.
Molecular Weight of Antigen 220 kDa
Purification Method Unpurified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5426
Test Specificity This is specific to the human DNA pol epsilon and does not recognize DNA pol alpha, beta or delta.
Target Species Human, Mouse, Mustelid, Primate, Drosophila (Negative)
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.