Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNA Polymerase epsilon catalytic subunit A, Mouse anti-Human,Mouse,Mustelid,Primate, Alexa Fluor 700, Clone: 3C5.1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100115AF7000.1ML
Description
DNA Polymerase epsilon catalytic subunit A Monoclonal specifically detects DNA Polymerase epsilon catalytic subunit A in Human, Mouse, Mustelid, Primate, Drosophila (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
DNA Polymerase epsilon catalytic subunit A | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
POLE | |
Human DNA Polymerase epsilon purified from HeLa cells. | |
0.1 mL | |
Chromatin Research, DNA Polymerases, DNA Repair | |
5426 | |
Human, Mouse, Mustelid, Primate, Drosophila (Negative) | |
Purified |
Western Blot, Immunofluorescence | |
3C5.1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
DKFZp434F222, DNA polymerase epsilon catalytic subunit A, DNA polymerase II subunit A, EC 2.7.7, EC 2.7.7.7, POLE1FLJ21434, polymerase (DNA directed), epsilon | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This is specific to the human DNA pol epsilon and does not recognize DNA pol alpha, beta or delta. | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction